Learn More
Thermo Scientific™ Thioglycollate Medium USP
Description
Cultivate aerobic and anaerobic organisms in the performance of sterility tests with Thermo Scientific™ Thioglycollate Medium USP prepared media tubes. The medium is prepared according to the formula specified in the US Pharmacopoeia1 for the performance of sterility tests, and conforms to formulations detailed in the British and European Pharmacopoeia 2,3.
Thioglycollate Medium USP is suitable for the cultivation of both aerobic and anaerobic organisms. No paraffin or special seal is necessary nor is an anaerobic jar required for the cultivation of anaerobes. It is well buffered, so that acid or alkaline inocula produce negligible alteration in the reaction of the medium. The sodium thioglycollate content of the medium will neutralize the bacteriostatic effect of mercurial compounds used as preservatives in solutions for injection, etc. If the solution undergoing test contains a bacteriostatic substance, it is necessary, in order to avoid a false negative result, to establish the bacteriostatic activity of the product by the method described in the US Pharmacopoeia1. Thioglycollate Medium USP is also recommended for the cultivation of Clostridium species.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Straw/yellow with or without red layer |
| Content And Storage | 2° to 25°C |
| Format | 125 mL Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Microbiology Media |
| Fill Weight | 100 to 101.5 g |
| Packaging Type | 10 Sirop Bottles with screw cap closures in a box |
| Quantity | 10/Pk. |
For In Vitro Diagnostic Use.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.