missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Sabouraud Dextrose Agar + Chloramphenicol Slope
Click to view available options
Fill Weight:
100 to 101.5 g
15-15.3 g
200 to 204 g
Format:
125 mL Sirop Bottle
250 mL Sirop Bottle
Universal bottle
Unit Size:
Pack of 10
Pack of 24
Description
Sabouraud Dextrose Agar and Chloramphenicol Slope (BO0422F) is an acidic pH medium for the detection of fungi and yeasts from clinical samples.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 25°C |
| Format | 250 mL Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 200 to 204 g |
| Packaging Type | 10 Sirop Bottles with Screw Cap Closures/Box |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction