Learn More
uvex X-Fit Guard Spectacles
Product Code. 17737699 Shop All Uvex Products
Change view
Click to view available options
Lens Tint: Clear
Product Code.
17737699 Lens Tint: Clear
1 options
Product Code. 17737699 Supplier uvex Supplier No. 9199180
  • ONLINE EXCLUSIVE PRICE
    £14.65
    £14.49 / Pack of 1
    Save £0.16
    1% Off
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
The Uvex X-Fit safety spectacles provide optimal eye coverage and high levels of wearer acceptance.
The uvex x-fit weighs just 23 grams. The x design of the lens and the translucent side arms make the uvex x-fit a real highlight.
General features
  • Classic, close fitting safety spectacle with extension kit options
  • Frameless lens design for a wide field of vision
  • Modular system allows functional accessories to be attached enhancing the ability to get on with the job
  • Robust polycarbonate lens with uvex’s proven supravision coating technology – permanent anti-fog and scratch-resistant lens
  • Clip-in frame seal provides extra protection against particles, dirt and dust
  • Metal-free
Safety features
  • Certified in line with EN 166 (personal protective eyewear) and EN 170 (UV protective filter)
  • Marking: W 166 FT CE – 2C-1,2 W1 FTKN CE
  • UV400 – 100% protection against dangerous UVA and UVB radiation up to 400 nm
Comfort features
  • Adjustment for every head and facial shape with the optional uvex Comfort Slider - a multi-stage temple inclination that allows wearers to achieve their own comfortable wearing position
  • Nose bridge designed for all-day comfort
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Chemical-resistant
Lens Coating Anti-fog, Scratch-resistant
Color Translucent, Gray
Certifications/Compliance (Europe) EN 166, EN 170
Lens Tint Clear
Product Type Ergonomic Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard