missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Antibody (rWT1/857), Novus Biologicals™
Mouse Monoclonal Antibody
£231.00 - £431.00
Specifications
| Antigen | WT1 |
|---|---|
| Clone | rWT1/857 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18246790
|
Novus Biologicals
NBP2-53181-20UG |
20 μg |
£231.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18147999
|
Novus Biologicals
NBP2-53181-100UG |
100 μg |
£431.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WT1 Monoclonal specifically detects WT1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| WT1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
| 1mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 7490 | |
| Recombinant full-length human WT1 protein | |
| Primary | |
| Store at 4C. |
| rWT1/857 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
| WT1 | |
| IgG1 κ | |
| Protein A or G purified | |
| 51 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title