missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Antibody (rWT1/857), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53181-100UG
This item is not returnable.
View return policy
Description
WT1 Monoclonal specifically detects WT1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| WT1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
| AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
| Mouse | |
| 51 kDa | |
| 100 μg | |
| Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
| 7490 | |
| Store at 4C. | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| rWT1/857 | |
| Unconjugated | |
| 1mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| WT1 | |
| Recombinant full-length human WT1 protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction