missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Wortmannin, Penicillium funiculosum, 99+%

Product Code. 15496119
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496119 10 mg 10mg
15406129 25 mg 25mg
15416129 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15496119 Supplier Thermo Scientific Chemicals Supplier No. J63983.MA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of phosphatidylinositol 3-kinase | CAS: 19545-26-7 | C23H24O8, C23H24O8 | 428.44 g/mol

Wortmannin is used as inhibitor of phosphatidylinositol 3-kinase, used cell biology reagent that has been used previously in research to inhibit DNA repair, receptor-mediated endocytosis and cell proliferation. It inhibits fMLP induced PIP3 and superoxide anion production in guinea pig neutrophils. It blocks insulin metabolic effects in rat adipocytes without affecting the insulin receptor tyrosine kinase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19545-26-7
Molecular Formula C23H24O8
Synonym KY 12420

Specifications

Quantity 10 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Inhibitor of phosphatidylinositol 3-kinase
Formula Weight 428.44g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.