missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Works Calibration Certificate - 5, 7 and 10% CO2

Factory calibration certificate for 5, 7 and 10% CO2

Brand:  Memmert™ ICO50D00106

Additional Details : Weight : 0.01100kg

 View more versions of this product

Product Code. 17120025

  • £290.00 / Each
Estimated Shipment: 20-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Special works calibration certificates upon request
  • For 5 %, 7 % and 10 % CO2 (measured at +37 °C)
Specifications

Specifications

ICO50
Works Calibration Certificate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers