missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Works Calibration Certificate - 5, 7 and 10% CO2
Factory calibration certificate for 5, 7 and 10% CO2
£290.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17120025
|
Memmert™
ICO50D00106 |
ICO50 |
£290.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17129924
|
Memmert™
ICO105D00106 |
ICO105 |
£290.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17159954
|
Memmert™
ICO150D00106 |
ICO150 |
£290.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17199984
|
Memmert™
ICO240D00106 |
ICO240 |
£290.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Special works calibration certificates upon request
- For 5 %, 7 % and 10 % CO2 (measured at +37 °C)
Specifications
Works Calibration Certificate |