missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT7B Monoclonal Antibody (C4), Invitrogen™

Mouse Monoclonal Antibody
£178.00 - £408.00
Specifications
| Antigen | WNT7B |
|---|---|
| Clone | C4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17604006
|
Invitrogen™
MA543544 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17693996
![]() |
Invitrogen™
MA543543 |
100 μL |
£408.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Wnt7b encodes a member of the WNT gene family, which consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Among members of the human WNT family, this gene product is most similar to WNT7A protein.Specifications
| WNT7B | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Protein Wnt-7b; wingless-related MMTV integration site 7B; wingless-type MMTV integration site family member 7B; wingless-type MMTV integration site family, member 7B; Wnt family member 7B; WNT7B; Wnt-7b | |
| Wnt7b | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| C4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P56706 | |
| 7477 | |
| Thr216-Lys349 (Accession P56706), with Two N-terminal Tags, His-tag and SUMO-tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Wnt7b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title