missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT7B Monoclonal Antibody (C4), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543543
This item is not returnable.
View return policy
Description
Wnt7b encodes a member of the WNT gene family, which consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Among members of the human WNT family, this gene product is most similar to WNT7A protein.
Specifications
| WNT7B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P56706 | |
| Wnt7b | |
| Thr216-Lys349 (Accession P56706), with Two N-terminal Tags, His-tag and SUMO-tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| C4 | |
| Unconjugated | |
| Wnt7b | |
| Protein Wnt-7b; wingless-related MMTV integration site 7B; wingless-type MMTV integration site family member 7B; wingless-type MMTV integration site family, member 7B; Wnt family member 7B; WNT7B; Wnt-7b | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 7477 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction