missing translation for 'onlineSavingsMsg'
Learn More

WNT2 Mouse anti-Human, Mouse, Rat, Clone: 3H7B10, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen WNT2
Clone 3H7B10
Concentration 1 mg/mL
Applications Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16888735
View Documents
Proteintech
66656-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16878735
View Documents
Proteintech
66656-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
Specifications

Specifications

WNT2
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Int 1 like protein 1, Int 1 related protein, INT1L1, IRP, Protein Wnt 2, WNT2
WNT2
IgG1
Protein G
Antibody
3H7B10
Immunofluorescence, Western Blot
Unconjugated
Mouse
Human, Mouse, Rat
P09544, P21552
114487, 22413, 7472
WNT2 Fusion Protein Ag25185
Primary
-20°C
WNT2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.