missing translation for 'onlineSavingsMsg'
Learn More

WNT2 Mouse anti-Human, Mouse, Rat, Clone: 3H7B10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66656-1-IG-150UL

 View more versions of this product

Product Code. 16878735

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
Specifications

Specifications

WNT2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P09544, P21552
WNT2
WNT2 Fusion Protein Ag25185
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunofluorescence, Western Blot
3H7B10
Unconjugated
WNT2
Int 1 like protein 1, Int 1 related protein, INT1L1, IRP, Protein Wnt 2, WNT2
Mouse
Protein G
RUO
114487, 22413, 7472
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.