missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-9b Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57937
This item is not returnable.
View return policy
Description
Wnt-9b Polyclonal specifically detects Wnt-9b in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Wnt-9b | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Protein Wnt-14b, wingless-type MMTV integration site family, member 15, wingless-type MMTV integration site family, member 9B, WNT14Bprotein Wnt-9b, WNT15Protein Wnt-15 | |
| Rabbit | |
| 39 kDa | |
| 100 μL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 7484 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| O14905 | |
| WNT9B | |
| Synthetic peptides corresponding to WNT9B (wingless-type MMTV integration site family, member 9B) The peptide sequence was selected from the C terminal of WNT9B. Peptide sequence FRETGQVLKLRYDSAVKVSSATNEALGRLELWAPARQGSLTKGLAPRSGD. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction