missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-3a Antibody (1E6G4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£198.00 - £431.00
Specifications
| Antigen | Wnt-3a |
|---|---|
| Clone | 1E6G4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18244090
|
Novus Biologicals
NBP2-52588 |
0.1 mg | |||||||
|
18602277
|
Novus Biologicals
NBP2-52588-0.025mg |
0.025 mg |
£198.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Wnt-3a Monoclonal specifically detects Wnt-3a in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Wnt-3a | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| 89780 | |
| Purified recombinant fragment of human WNT3A (AA: 170-352) expressed in E. Coli. | |
| Primary | |
| 39.4 kDa |
| 1E6G4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A | |
| WNT3A | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title