missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-3a Antibody (1E6G4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52588
This item is not returnable.
View return policy
Description
Wnt-3a Monoclonal specifically detects Wnt-3a in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Wnt-3a | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A | |
| Mouse | |
| 39.4 kDa | |
| 0.1 mg | |
| Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 89780 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 1E6G4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| WNT3A | |
| Purified recombinant fragment of human WNT3A (AA: 170-352) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction