missing translation for 'onlineSavingsMsg'
Learn More

Wnt-2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £393.00

Specifications

Antigen Wnt-2
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18636012
View Documents
Novus Biologicals
NBP2-94468-0.02ml
0.02 mL
£169.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18636022
View Documents
Novus Biologicals
NBP2-94468-0.1ml
0.1 mL
£393.00
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Wnt-2 Polyclonal antibody specifically detects Wnt-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Wnt-2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
PBS (pH 7.3), 50% glycerol
7472
IgG
Affinity purified
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human, Mouse, Rat
INT1L1secreted growth factor, Int-1-like protein 1, Int-1-related protein, IRPprotein Wnt-2, wingless-type MMTV integration site family member 2
Recombinant fusion protein containing a sequence corresponding to amino acids 26-360 of human WNT2 (NP_003382.1). SWWYMRATGGSSRVMCDNVPGLVSSQRQLCHRHPDVMRAISQGVAEWTAECQHQFRQHRWNCNTLDRDHSLFGRVLLRSSRESAFVYAISSAGVVFAITRACSQGEVKSCSCDPKKMGSAKDSKGIFDWGGCSDNIDYGIKFARAFVDAKERKGKDARALMNLHNNRAGRKAVKRFLKQECKCHGVSGSCTLRTCWLAMADFRKTGDYLWRKYNGAIQVVMNQDGTGFTVANERFKKPTKNDLVYFENSPDYCIRDREAGSLGTAGRVCNLTSRGMDSCEVMCCGRGYDTSHVTRMTKCGCKFHWCCAVRCQDCLEALDVHTCKAPKNADWTTAT
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.