missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94468-0.02ml
This item is not returnable.
View return policy
Description
Wnt-2 Polyclonal antibody specifically detects Wnt-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Wnt-2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| INT1L1secreted growth factor, Int-1-like protein 1, Int-1-related protein, IRPprotein Wnt-2, wingless-type MMTV integration site family member 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 26-360 of human WNT2 (NP_003382.1). SWWYMRATGGSSRVMCDNVPGLVSSQRQLCHRHPDVMRAISQGVAEWTAECQHQFRQHRWNCNTLDRDHSLFGRVLLRSSRESAFVYAISSAGVVFAITRACSQGEVKSCSCDPKKMGSAKDSKGIFDWGGCSDNIDYGIKFARAFVDAKERKGKDARALMNLHNNRAGRKAVKRFLKQECKCHGVSGSCTLRTCWLAMADFRKTGDYLWRKYNGAIQVVMNQDGTGFTVANERFKKPTKNDLVYFENSPDYCIRDREAGSLGTAGRVCNLTSRGMDSCEVMCCGRGYDTSHVTRMTKCGCKFHWCCAVRCQDCLEALDVHTCKAPKNADWTTAT | |
| 0.02 mL | |
| Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 7472 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction