missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WISP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £386.00
Specifications
| Antigen | WISP2 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18675011
|
Novus Biologicals
NBP2-93612-0.02ml |
0.02 mL |
£171.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603760
|
Novus Biologicals
NBP2-93612-0.1ml |
0.1 mL |
£386.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WISP2 Polyclonal antibody specifically detects WISP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| WISP2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Stem Cells | |
| PBS (pH 7.3), 50% glycerol | |
| 8839 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Connective tissue growth factor-like protein, Connective tissue growth factor-related protein 58, CT58CTGF-Lwnt-1 signaling pathway protein 2, CTGFL, WISP-2, WNT1 inducible signaling pathway protein 2, WNT1-inducible-signaling pathway protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 164-250 of human WISP2 (NP_003872.1). GQGGGLGTQPLPAQGPQFSGLVSSLPPGVPCPEWSTAWGPCSTTCGLGMATRVSNQNRFCRLETQRRLCLSRPCPPSRGRSPQNSAF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title