missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, Protect coated, GLS 80™ Thread, Plastic safety coated, Graduated

Product Code. 10295203 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
3500 mL
5000 mL
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10295203 5000 mL Each
11796988 500 mL Pack of 10
11706998 1000 mL Pack of 10
11716998 2000 mL Pack of 10
12666167 250 mL Pack of 10
11619938 3500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10295203 Supplier DWK Life Sciences Supplier No. 1160165

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Wide mouth, multifunctional glass laboratory bottles with protective safety coating.

DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside. The bottles are moulded from high-purity, clear, glass that is then coated with a clear PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134°C.

  • The coating provides scratch, leak* and splinter* protection (* only applies to bottles 5000 ml and less)
  • UV protection up to approx. 380 nm due to external coating
  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • Special thread design for rapid opening and closing in less than a turn
  • Optimum filling and pouring and particularly easy cleaning
  • Spoons, spatulas and tweezers can be manipulated with ease
  • Even large-volume funnels can be inserted into the bottles without any difficulty
  • Stability due to the large base area
  • Easy-to-read scale with graduation marks to estimate the content
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5000 mL
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Includes With screw cap and pouring ring from PP (blue)
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Thread Size (Metric) GLS 80
Closure Material Polypropylene
Diameter (Metric) 182 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.