missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Reagent Bottle, Wide Neck, Clear

Product Code. 11705374 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
250 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11705374 50 mL Pack of 10
11736133 250 mL Pack of 10
11776253 100 mL Pack of 10
11786253 500 mL Pack of 10
11796253 1000 mL Pack of 10
15281659 2000 mL Pack of 10
10393921 5000 mL Each
10559251 10,000 mL Each
10268800 20,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11705374 Supplier DWK Life Sciences Supplier No. 211851707

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Classic wide-neck, glass-stoppered reagent bottles.

The classical stoppered reagent bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a wide conical ground joint neck and comply with ISO 4796-2. The wider neck is more convenient for handling powders. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel.

  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • All borosilicate glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to standard ground joint
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic print
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 50 mL
Description Neck with standard ground joint and glass flat-headed stopper
For Use With (Application) Storage of air sensitive reagents, pharmaceuticals, and chemicals; Storage of hygroscopic liquids and pharmaceuticals; Transport and shipping of samples
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Diameter (Metric) 44 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.