missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, GLS 80™ Thread, Graduated, with Screw Cap and Pouring Ring

Product Code. 10318322 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
3500 mL
5000 mL
10 L
20 L
30 L
50 L
Unit Size:
Each
Pack of 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10318322 10 L Each
10256272 500 mL Pack of 10
10378322 1000 mL Pack of 10
10563253 2000 mL Pack of 10
10114642 5000 mL Each
10113772 20 L Each
11699928 3500 mL Each
13127236 250 mL Pack of 10
11301465 50 L Each
11311465 30 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 10318322 Supplier DWK Life Sciences Supplier No. 1113950
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Wide mouth, multifunctional glass laboratory bottles.

DURAN™ wide mouth GLS 80™ laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ wide mouth GLS 80™ are supplied complete with blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.

  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • Special thread design for rapid opening and closing in less than a turn
  • Optimum filling and pouring and particularly easy cleaning
  • Spoons, spatulas and tweezers can be manipulated with ease
  • Even large-volume funnels can be inserted into the bottles without any difficulty
  • Stability due to the large base area
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
  • Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10 L
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Includes Blue Polypropylene Screw Cap and Pouring Ring
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Thread Size (Metric) GLS 80
Closure Material Polypropylene
Diameter (Metric) 227 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.