Learn More
Fisherbrand™ White Polypropylene Caps
Product Code. 16478125 Shop All Fisherbrand Products
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Liner Material:
PTFE/Polyethylene Foam
Polyethylene Foam
Pulp/Vinyl
Quantity:
100/Cs.
1300/Cs.
144/Cs.
2100/Cs.
288/Cs.
370/Cs.
500/Cs.
Unit Size:
Pack of 100
Pack of 1300
Pack of 144
Pack of 2100
Pack of 288
Pack of 370
Pack of 500
Product Code. 16478125
Only null left
Add to basket
Add to basket
White polypropylene screw caps provides clear surface for writing on cap
  • Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
  • Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Color White
Material Polypropylene
Quantity 1300/Cs.
Liner Material Polyethylene Foam
Neck Finish 53-400
Product Type Lined Cap