Learn More
Fisherbrand™ White Polypropylene Caps
Shop All Fisherbrand Products
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Liner Material:
PTFE/Polyethylene Foam
Polyethylene Foam
Pulp/Vinyl
Quantity:
100/Cs.
1300/Cs.
144/Cs.
2100/Cs.
288/Cs.
370/Cs.
500/Cs.
Unit Size:
Pack of 100
Pack of 1300
Pack of 144
Pack of 2100
Pack of 288
Pack of 370
Pack of 500
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| Quantity | 100/Cs. |
| Liner Material | PTFE/Polyethylene Foam |
| Neck Finish | 58-400 |
| Product Type | Lined Cap |