missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ White Polypropylene PE Foam/PTFE Cap

Product Code. 15593261 Shop All Fisherbrand Products
Change view
Click to view available options
Neck Finish:
20-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Quantity:
100/Pk.
144/Cs.
288/Pk.
500/Cs.
Unit Size:
Pack of 144
Pack of 288
Pack of 500
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Quantity unitSize
15593261 20-400 100/Pk. Pack of 500
15503271 24-400 100/Pk. Pack of 500
15513271 28-400 100/Pk. Pack of 500
15523271 33-400 100/Pk. Pack of 500
11528192 38-400 500/Cs. Pack of 500
11538192 43-400 500/Cs. Pack of 500
11548192 45-400 500/Cs. Pack of 500
15533271 48-400 100/Pk. Pack of 500
15543271 53-400 100/Pk. Pack of 500
15553271 58-400 100/Pk. Pack of 500
11598192 70-400 144/Cs. Pack of 144
15563271 83-400 144/Cs. Pack of 144
11518202 89-400 144/Cs. Pack of 144
11588192 63-400 288/Pk. Pack of 288
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 15593261

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene PE Foam/PTFE Cap have a PTFE facing, backed by F217 liners.

Polypropylene (PP) caps with F217 & PTFE liners are an economical alternative to Thermoset & Phenolic PTFE lined caps. They have a PTFE facing, backed by F217 liners. The F217 liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE). PTFE/F217 liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference.

  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference

Specifications

Color White
Description Screw cap
Material Polypropylene
No. per Case 100/Pk.
Packaging Packed in 5 bags of 100
Quantity 100/Pk.
Sterility Sterile
Liner Material PE Foam/PTFE
Neck Finish 20-400
Certifications/Compliance ISO 9001
Shape Round
Product Type PE Foam/PTFE-lined Cap
For Use With (Equipment) Bottle
Closure Material Polypropylene
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.