missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ White Dosing Spoon
GREENER_CHOICEBIOPHARMA

Product Code. 16555220 Shop All Bürkle Products
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
0.5 mL
1 mL
2.5 mL
5 mL
10 mL
15 mL
25 mL
50 mL
Sterility:
Non-sterile
Sterile
missing translation for 'unitSize'
Pack of 100
This item is not returnable. View return policy

Product Code. 16555220

missing translation for 'mfr': Buerkle™ 53784007

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Volumetric and dosing spoons can be used to precisely measure small amounts of powder, granules, and liquids.

  • The content volume is engraved on the handle
  • The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, allowing for good emptying of small remaining amounts
  • The flat base makes it possible to set the volumetric spoon down without spilling the content
  • The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities
  • Since the plastic volumetric spoon was developed for single use, there is no cleaning and the possibility of cross-contamination and impurities is excluded
  • The dosing spoons and volumetric spoons are suitable for precise measuring for chemical and pharmaceutical applications, cosmetics, animal feed, and the food and beverage industry
  • Sterile versions are individually packaged
  • With stripper edge
  • For single use
  • Cleanroom manufactured (Class 7)
  • Complies with EU food requirements and FDA regulations
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100/Pk.
Description Dosing spoons with stripper edge for exact measuring
Sterility Sterile
Capacity (Metric) 1 mL
Color White
Certifications/Compliance Food Contact, Reach, RoHS
Disposable Single-use
Product Type Spoon
For Use With (Application) Especially for The Food, Animal Feed and Pharmaceutical Industries
Material Polystyrene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.