Learn More
Coverguard™ White Coverall 5M20
Product Code. 16302068
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16302068 Large Pack of 25
16382058 Small Pack of 25
16392058 Medium Pack of 25
16312068 X-Large Pack of 25
16322068 2X-Large Pack of 25
16332068 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16302068 Supplier Coverguard™ Supplier No. 44303
Only null left
Add to basket
Add to basket
Protective coveralls made from laminated microporous films are effective against particles and splashes of non-hazardous chemical liquids. The evaporation of water vapor allows for greater wearing comfort. Elasticated hood, waist, wrists and ankles prevent the garment riding up during movement.
  • CE PPE Cat. III
  • Limited protection against chemical liquid splash
  • Protects against chemical solid particles in the air
  • Anti-static protection
  • White
  • 55 g/m2 SPP + microporous film fabric
  • Central zip with adhesive flap
  • 3 pans elasticated hood
  • Elasticated: waist, wrist with elastic thumbloop, ankle tightening, slingbar
  • Self-adhesive chin-strap
Specifications
Size Large
Gender Unisex
Material PPE
Color White
Closure Type Central Zip with Adhesive Flap
Sterility Non-sterile
Coating Microporous Film
Back Style Elasticated
Certifications/Compliance (Europe) CE PPE Cat. III, EN 13034:2005 + A1:2009 Type 6; EN ISO 13982-1:2004 + A1:2020 Type 5; EN 1149-5:2008