Learn More
Fisherbrand™ Whirlimixer Plus Platform Mixers
Product Code. p-8013197 Shop All Fisherbrand Products
Click to view available options
For Use With (Application):
For 1.5 mL Micro Tubes
For Microtitre Plate
Unit Size:
Each
Product Code. 10164243
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for tube, microplate, flask and microtube mixing using accessory platforms which are quickly interchangeable.
  • For tube, microplate, flask and microtube mixing using accessory platforms which are quickly interchangeable.
  • Variable speed control giving controlled mixing over a speed range
  • Where accessory platforms are fitted, the unit can operate at speeds for gentle mixing reducing damage to cell structures
  • Two modes of operation - set for continuous running or press-start
  • Operation suitable for use with accessory tube mixing cup
  • Accessory platforms are autoclavable
Specifications
Product Type Mixer Platform
Model Whirlimixer Plus
For Use With (Application) For Microtitre Plate