Learn More
Fisherbrand™ Whirlimixer Plus Platform Mixers
Shop All Fisherbrand Products
Click to view available options
For Use With (Application):
For 1.5 mL Micro Tubes
For Microtitre Plate
Unit Size:
Each
Description
- For tube, microplate, flask and microtube mixing using accessory platforms which are quickly interchangeable.
- Variable speed control giving controlled mixing over a speed range
- Where accessory platforms are fitted, the unit can operate at speeds for gentle mixing reducing damage to cell structures
- Two modes of operation - set for continuous running or press-start
- Operation suitable for use with accessory tube mixing cup
- Accessory platforms are autoclavable
Specifications
Specifications
| Product Type | Mixer Platform |
| Model | Whirlimixer Plus |
| For Use With (Application) | For Microtitre Plate |