Learn More
DWK Life Sciences Wheaton™ Polyacetal Clamps
Color-coded clamps hold standard taper joints securely under moderate pressure
Brand:  DWK Life Sciences 297749
Additional Details : Weight : 0.47630kg
 View more versions of this product
Product Code. 12448922
  • £217.00 / Pack of 12
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
  • Molded from polyoxymethylene (POM), an acetal resin
  • Will not scratch glass and can withstand temperatures up to 150°C
  • Resistant to bases and weak acids
  • Not resistant to halogen gases and concentrated acids
Specifications
Specifications
Standard Taper Clamp
POM Acetal
Kuderna Danish Evaporative Concentrator
Blue