Learn More
DWK Life Sciences Wheaton™ Polyacetal Clamps
Color-coded clamps hold standard taper joints securely under moderate pressure
£209.00 - £225.00
Specifications
Type Standard Taper Clamp
Material POM Acetal
View More Specs
Products 3
Product Code Brand Color For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Color For Use With (Equipment) Price Quantity & Availability  
12438922
View Documents Promotion Details
DWK Life Sciences
297746
Yellow Kuderna Danish Evaporative Concentrator
£209.00
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
12448922
View Documents Promotion Details
DWK Life Sciences
297749
Blue Kuderna Danish Evaporative Concentrator
£217.00
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
12458922
View Documents Promotion Details
DWK Life Sciences
297752
Dark Green Wheaton Sulfide Distillation Kit
£225.00
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • Molded from polyoxymethylene (POM), an acetal resin
  • Will not scratch glass and can withstand temperatures up to 150°C
  • Resistant to bases and weak acids
  • Not resistant to halogen gases and concentrated acids
Specifications
Specifications
Standard Taper Clamp
POM Acetal