missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Low-Profile Reservoirs

Replacement glass reservoirs for Acurex™ Low-Profile Dispensers

Brand:  DWK Life Sciences 844082

Additional Details : Weight : 0.27220kg

 View more versions of this product

Product Code. 12447201

  • £50.10 / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket

Description

Description

  • Replacement 250mL Amber Glass Reservoir
Specifications

Specifications

Reservoir
Glass
Acurex™ low profile dispenser
Amber
250 mL
8.45 oz.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers