missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper

Product Code. 12472481 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Unit Size:
Pack of 10
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12472481 250 mL Pack of 10
12349719 100 mL Pack of 6
12482481 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12472481 Supplier DWK Life Sciences Supplier No. 215257
  • ONLINE EXCLUSIVE PRICE
    £551.25
    £540.10 / Pack of 10
    Save £11.15
    2% Off
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for shelf storage

  • Amber borosilicate glass bottle with narrow mouth
  • Protects light-sensitive reagents and solutions
  • Ground-glass octagon flat stopper provides excellent seal; protects top lip of bottle from dust

Specifications

Capacity (Metric) 250 mL
Color Amber
Diameter (English) 2.83 in.
Material Glass
Closure Color Amber
Mouth Narrow
Height (Metric) 145 mm
Height (English) 5.7 in.
Closure Material Borosilicate Glass
Diameter (Metric) 72 mm
Capacity (English) 8.45 oz.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.