missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ UniFilter™ 384-Well Microplates
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
£1515.00 - £1945.00
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Color | Clear |
Material | Polystyrene |
No. of Wells | 384 |
Product Code | Brand | Membrane | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Membrane | For Use With (Application) | Price | Quantity & Availability | ||||
11304884
|
Cytiva
7700-1101 |
Whatman GF/C | Can be used with gel filtration media for high-throughput sequencing reaction clean-up, including removal of dye blobs |
£1515.00
Pack of 50 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
11988975
|
Cytiva
7700-2106 |
0.45 μm Hydrophilic PVDF | Used for DNA template clean-up, cell capture, and for the removal of unwanted debris |
£1815.00
Pack of 50 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11314884
|
Cytiva
7700-2110 |
DNA Binding | Can be used with gel filtration media for high-throughput sequencing reaction clean-up, including removal of dye blobs |
£1945.00
Pack of 50 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
Description
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Specifications
Standard | |
Clear | |
384 |
16 x 24 | |
Polystyrene | |
Long drip director |