Learn More
Cytiva Whatman™ UniFilter™ 384-Well Microplates
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Brand: Cytiva 7700-1101
Code : A0
Additional Details : Weight : 0.38000kg
Description
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Specifications
Standard | |
GE’s Whatman UNIFILTER 384 well microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples. | |
Clear | |
384 | |
Can be used with gel filtration media for high-throughput sequencing reaction clean-up, including removal of dye blobs | |
50/Pk. |
Whatman GF/C | |
16 x 24 | |
Polystyrene | |
Filter | |
Long drip director |
Safety and Handling
- FILTER GLASS CIRCLE
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : glass, oxide, chemicals
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.