Learn More
Cytiva Whatman™ 30mm Puradisc GF 92 Syringe Filters
Designed to be resistant to organic solvents. Whatman™ 30mm Puradisc GF 92 Syringe Filters are highly suitable for clarification of aggressive organic solvents.
Brand: Cytiva 10463543
Code : A0
Additional Details : Weight : 0.43000kg
Description
- Glass fiber is ideal for prefiltration of high solid content solutions
- Use as a venting filter for small vessels
- Extremely high chemical resistance
- Permits optimal utilization of small sample volumes
Specifications
Glass Fiber 92 | |
Polypropylene | |
100 | |
Syringe Filter |
>1 μm | |
Female luer lock | |
Male luer lock | |
30 mm |
Safety and Handling
- FILTER GLASS CIRCLE
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : glass, oxide, chemicals
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.