missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ MD050 and MD142 Series Pressure Filtration Manifolds with Infusion Cylinders
Cytiva Pressure Filtration Manifolds come in PTFE or stainless steel. Equipped with infusion cylinders, they are suitable for batch filtration from 20 mL, filtration of liquids/gases, ‘sterile’ filtration of serum, or clear filtration of viscous media using membranes, paper, or glass fiber discs.
£1495.00 - £4635.00
Specifications
Description | Pressure filtration device |
---|---|
Disposable | Reusable |
Sterility | Non-sterile |
For Use With (Application) | Pressure Filtration |
Temperature (English) Max. | 392°F |
Product Code | Brand | Model | Material | Volume (Metric) | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Material | Volume (Metric) | Includes | Price | Quantity & Availability | ||
11335004
|
Cytiva
10450450 |
MD050/4 | Stainless Steel | 200 mL | Silicone O-Ring/PTFE O-Ring, 200 mL Infusion Cylinder for 50 mm Membrane, Rapid Seal Connector |
£1495.00
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
11365004
|
Cytiva
10451610 |
MD142/5/3 | Stainless Steel | 2200 mL | Silicone O-Ring/PTFE O-Ring, 2200 mL Infusion Cylinder for 142 mm Membrane |
£4600.00
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
11395004
|
Cytiva
10451710 |
MD142/7/3 | PTFE | 1500 mL | PTFE O-Ring, 1500 mL Infusion Cylinder for 142 mm Membrane |
£4635.00
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||
Description
- Product Type: Pressure Filtration Manifold
- Includes: Infusion Cylinder
- Temperature Resistance Maximum: 200°C
Specifications
Pressure filtration device | |
Non-sterile | |
392°F | |
Pressure Filtration Manifold |
Reusable | |
Pressure Filtration | |
200°C |