missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polydisc In-line Filter
For serum samples
£165.12 - £525.64
Product Code | Brand | Material | Pore Size | Flow Rate | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Pore Size | Flow Rate | Quantity | Price | Quantity & Availability | ||
11319204
|
Cytiva
6724-5000 |
PES | 1.0 μm | 500.0mL/min./cm sq. | 10/Pk. |
£168.00
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||
11304644
|
Cytiva
6724-5002 |
PES | 0.2 μm | 500.0mL/min./cm sq. | 10/Pk. |
£165.12
Pack of 10 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
||
11399194
|
Cytiva
2227 |
Polypropylene | 5 μm | 1500mL/min | 50 |
£525.64
Pack of 50 |
Estimated Shipment: 06-12-2024
Log in to see stock. |
||
Description
- Glass microfiber and polyethersulfone membranes
- Contains stack of filter media
- Radiation sterilized