missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polydisc In-line Filter

For serum samples

Brand:  Cytiva 2227

Code : A0

Additional Details : Weight : 0.77000kg

 View more versions of this product

Product Code. 11399194

  • £525.64 / Pack of 50
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

  • Glass microfiber and polyethersulfone membranes
  • Contains stack of filter media
  • Radiation sterilized

Specifications

Specifications

Polypropylene
1500mL/min
50
5 μm
For large volume sample preparation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers