missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Grade 91 Wet Strengthened Qualitative Filter Papers - Sheets
High wet strength due to the addition of chemically stable resin
£940.00 - £970.00
Specifications
Format | Sheet |
---|---|
Material | Cellulose |
Pore Size | 10 μm |
Filtration Speed | 70 sec. |
For Use With (Application) | A general purpose creped filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration. |
Product Code | Brand | Dimensions (L x W) | Length (Metric) | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Length (Metric) | Width (Metric) | Price | Quantity & Availability | |||
11300534
|
Cytiva
1091-930 |
58 x 58 cm | 580 mm | 580 mm |
£940.00
Pack of 500 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||
11310534
|
Cytiva
1091-935 |
61 x 61 cm | 610 mm | 610 mm |
£970.00
Pack of 500 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||
Description
- Wet strengthened grade
- General purpose creped filter
- For less critical routine analysis
- Used to assay sucrose in cane sugar
Specifications
Sheet | |
10 μm | |
A general purpose creped filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration. | |
500 Pieces |
Cellulose | |
70 sec. | |
Grade 91 | |
205 μm |