missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Grade 91 Wet Strengthened Qualitative Filter Papers - Sheets
High wet strength due to the addition of chemically stable resin
Brand: Cytiva 1091-935
Code : 69
Additional Details : Weight : 11.40000kg
Description
- Wet strengthened grade
- General purpose creped filter
- For less critical routine analysis
- Used to assay sucrose in cane sugar
Specifications
Grade 91 Qualitative Filter Paper Wet-Strengthened, sheet, 61 x 61 cm | |
Cellulose | |
70 sec. | |
Grade 91 | |
205 μm | |
610 mm |
Sheet | |
10 μm | |
A general purpose creped filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration. | |
500 Pieces | |
61 x 61 cm | |
610 mm |