missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Grade 470
For electrophoresis and amino acid chromatography
£48.80 - £625.00
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
11338944
|
Cytiva
10318493 |
460 mm | 570 mm |
£625.00
Pack of 100 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||||
11328944
|
Cytiva
10318489 |
203 mm | 305 mm |
£48.80
Pack of 25 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
13464547
|
Cytiva
10539028 |
137.1m | 38 mm |
£118.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||||
Description
- Soft surface
- For gelatinous samples
- For the absorption of culture media
- Used as blotting paper and amino acid chromatography
Specifications
470 |