missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences WET FRED Set
The WET FRED Set is an application aid for large volumes of lysates or culture supernatants
£113.00 - £130.00
Specifications
Quantity | 1 Set |
---|---|
For Use With (Application) | Eases Protein Purification |
Type | Protein Purification Aid |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15583067
|
IBA Lifesciences
2-0911-001 |
1 mL Gravity Flow Columns |
£113.00
1 set |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
15573067
|
IBA Lifesciences
2-0910-001 |
5 to 10 mL Gravity Flow Columns |
£130.00
1 set |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
Description
- WET FRED enables the application of large cell culture supernatant or lysate volumes to a gravity flow column in a simple way
- Based on the siphon principle, it connects the gravity flow column with the bottle containing culture supernatant, lysate, or buffer that are to be applied
Specifications
1 Set | |
Eases Protein Purification | |
Protein Purification Aid |
For Research Use Only.