missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ West Nile Virus Envelope (lineage 1, strain NY99) Recombinant Rabbit Monoclonal Antibody (087)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£132.00 - £357.00

Specifications

Antigen West Nile Virus Envelope (lineage 1, strain NY99)
Clone 087
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30535136
GREENER_CHOICE
View Documents
Invitrogen™
MA560367
100 μL
£357.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30535461
View Documents
Invitrogen™
MA560368
20 μL
£132.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

West Nile Virus Envelope (lineage 1, strain NY99) Recombinant Monoclonal Antibody for Western Blot

West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins which play a major role for WNV entry into target cells. The viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway as delivery of core gene delivery into the striatum of mouse brain and skeletal muscle resulted in cell death and inflammation.
Specifications

Specifications

West Nile Virus Envelope (lineage 1, strain NY99)
1 mg/mL
Western Blot
Unconjugated
Rabbit
RUO
PBS with no preservative
Recombinant West Nile Virus (WNV) (lineage 1, strain NY99) E/Envelope Protein (Domain III) protein (Gln586-Ala705).
Antibody
087
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Virus
West Nile envelope; West Nile Virus; WNV envelope
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.