missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus C Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | West Nile Virus C Protein |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | ELISA 1 μg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
Description
West Nile Virus C Protein Polyclonal antibody specifically detects West Nile Virus C Protein in Virus samples. It is validated for ELISA.Specifications
| West Nile Virus C Protein | |
| ELISA 1 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| NP_776011 | |
| 912267 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Capside protein C, Core protein, West Nile Core, West Nile Virus Core protein, WNV core | |
| Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the amino-terminus of the WNV Core protein. The immunogen is located within the first 50 amino acids of WNV Core . Amino Acid Squence: SKKPGGPGKNRAVNM | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title