missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus C Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76435-0.025mg
This item is not returnable.
View return policy
Description
West Nile Virus C Protein Polyclonal antibody specifically detects West Nile Virus C Protein in Virus samples. It is validated for ELISA.
Specifications
| West Nile Virus C Protein | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Capside protein C, Core protein, West Nile Core, West Nile Virus Core protein, WNV core | |
| Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the amino-terminus of the WNV Core protein. The immunogen is located within the first 50 amino acids of WNV Core . Amino Acid Squence: SKKPGGPGKNRAVNM | |
| 0.025 mg | |
| Primary | |
| Virus | |
| IgG |
| ELISA | |
| 1 mg/mL | |
| ELISA 1 μg/mL | |
| NP_776011 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 912267 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction