missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Wellwash™ Microplate Washer (EU IVD/CE-marked)

Product Code. 15311095 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
Wellwash 1x12 IVD model (Not available in USA)
Wellwash 1x8 IVD model (Not available in USA)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
15311095 Wellwash 1x12 IVD model (Not available in USA) Each
15301095 Wellwash 1x8 IVD model (Not available in USA) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Fulfills IVD Directive 98/79/EC requirements, valid only in Europe. Not for in vitro diagnostics use in North America.
Product Code. 15311095 Supplier Thermo Scientific™ Supplier No. 5165080

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Fulfills IVD Directive 98/79/EC requirements, valid only in Europe. Not for in vitro diagnostics use in North America.

Wash 96-well plates securely with this easy-to-use, reliable microplate-strip washer for routine ELISA applications.

Wash 96-well plates securely with the Thermo Scientific™ Wellwash™ Microplate Washer, an easy-to-use, reliable microplate-strip washer for routine ELISA applications. The Wellwash features a graphical user interface, local language versions and USB port for optimal ease-of-use and convenience.

Simplify your ELISA workflow by using our eGuide to see our complete portfolio for all your ELISA needs.

Features:

  • Large color clear display and keypad for easy, convenient use
  • User interface is logic and intuitive for quick protocols development
  • Easy-to-use software requires minimal training for fast startup
  • Unpressurized bottles are safe and secure to use
  • Liquid level sensors are present in both wash and waste bottle for security
  • Automatic prime feature assure safe performance
  • Aerosol cover prevents aerosolization of infectous diseases
  • Plate sensor recognizes if plate is present
  • Wellwash is designed to efficiently remove the excess liquid from the wells
  • Low residual volumes provide optimal washing performance and reliable assay results

Includes:

1x2L wash bottle, 1x2L waste bottle, 1x8L wash head, aerosol cover

Recommended for:

ELISA-based assays

Warranty: 1 year

TRUSTED_SUSTAINABILITY

Specifications

Accuracy < 5% @ 300μL
Certifications/Compliance CE Marked
Connections USB
Wash/Waste Bottle Capacity 1
Bottles Wash Bottle (2 L), Waste Bottle (2 L)
Precision < 3% (CV) @ 300μL (96-well plate)
Dispense Volume 50 to 400 μL
Wash Heads 1 x 12
Plate Type 96-well Plate
Residual Volume <2 μL
Weight (Metric) 8 kg
Wash Volume 50 to 1000 μL
Display LCD
For Use With (Application) ELISA-Based Assays
Frequency 50/60 Hz
Model Wellwash 1x12 IVD model (Not available in USA)
Cell Washing No
Includes 1 x 12 wash head, 1 x 2L wash bottle, 1 x 2L waste bottle, aerosol cover
Width (English) 15.2 in.
Width (Metric) 385 mm
Height (English) 13.6 in.
Height (Metric) 345 cm
Depth (English) 9.4 in.
Depth (Metric) 240 mm
Automation Interface No
Weight (English) 17.6 lb.
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Prime Volume 5 to 100 mL
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Fulfills IVD Directive 98/79/EC requirements, valid only in Europe. Not for in vitro diagnostics use in North America.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.