missing translation for 'onlineSavingsMsg'
Learn More

Welch™ Gemini™ Dry Vacuum Pump: Model 2060

Economica, go-anywherel pumps with advanced chemical resistance

Brand:  Welch™ 2060C-02

 View more versions of this product

Product Code. 11786099

Alert:

Not recommended for use with strong acids or bases. For organic fume applications, check for chemical compatibility with Viton™. Not recommended for rotary evaporator, concentrator or vacuum oven applications.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.


Includes: AC/DC adapter, car adapter
This item is not returnable. View return policy

Description

Description

Ideal for organic filtration/degassing applications using aqueous solutions, select solvents, and moderate pH acids and bases. High-performance wetted components will not corrode. Use in lab or field.

  • Lightweight, compact and portable
  • Oil-free
  • Chemically resistant wetted parts (PARA heads and Viton™ diaphragms, tubing and valves)

Includes regulator and dial vacuum gauge for fine vacuum adjustment

Filtration, aspiration, degassing, and desiccation.

Order Info

DC car adapter is included for field use (Part No. 1431B).

Specifications

Specifications

0.008 hp
5.9 kg
230 V 50 Hz
European (Schuko)
4.98 kg
50 Hz
21.6 cm
23.5 cm
266 mbar
13 lb.
AC/DC adapter, regulator and dial vacuum gauge
2060
11 lb.
13 L/min.
8.5 in.
9.25 in.
7.25 in.
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers