missing translation for 'onlineSavingsMsg'
Learn More

Welch™ Vacuum Pump On/Off Foot Switch

Product Code. 10349442 Shop All Welch Ilmvac Products
Change view
Click to view available options
Electrical Requirements:
90 to 230 V
Model:
Momentary On/Off switch logic
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Model unitSize
10349442 90 to 230 V Momentary On/Off switch logic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10349442 Supplier Welch™ Supplier No. 1430C

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Vacuum pump accessory that provides hands-free operation of Welch DryFast diaphragm and WOB-L piston pumps.

  • Hands-free on/off pump operation
  • No adaptors or special connection needed
  • Plugs directly into the power source outlet
  • Switch logic and power varies depending on model:
    • Parts 01-104-17 and 01-055-16: 3-prong U.S. standard plug with 8 ft. cord (CSA, NEMA, and UL Enclosure Type 1)
    • Part 01-055-17: Male and female IEC connections with 8 ft. cord (plugs directly into the IEC connection on the pump)
TRUSTED_SUSTAINABILITY

Specifications

Electrical Requirements 90 to 230 V
For Use With WOB-L piston pumps
Model Momentary On/Off switch logic
Type Foot Switch
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.