Learn More
Invitrogen™ WDSOF1 Recombinant Rabbit Monoclonal Antibody (24GB3745)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557011
 View more versions of this product
Product Code. 30275456
  • £434.00 / 100µL
Description
WDSOF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Possible role in ribosomal RNA processing. May function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
WDSOF1
Recombinant Monoclonal
Unconjugated
DCAF13
Dcaf13; DDB1 and CUL4 associated factor 13; DDB1- and CUL4-associated factor 13; Gm83; HSPC064; WD repeat and SOF domain-containing protein 1; WD repeats and SOF domain containing 1; WD repeats and SOF1 domain containing; WDSOF1
Rabbit
Affinity chromatography
RUO
223499, 25879, 362902
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3745
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6PAC3, Q9NV06
DCAF13
A synthesized peptide derived from human DCAF13 (aa 1-46).
100 μL
Primary
Human, Mouse, Rat