missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDSOF1 Recombinant Rabbit Monoclonal Antibody (24GB3745)

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen WDSOF1
Clone 24GB3745
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275456
View Documents
Invitrogen™
MA557011
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275670
View Documents
Invitrogen™
MA557012
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

WDSOF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Possible role in ribosomal RNA processing. May function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.
Specifications

Specifications

WDSOF1
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
DCAF13
Dcaf13; DDB1 and CUL4 associated factor 13; DDB1- and CUL4-associated factor 13; Gm83; HSPC064; WD repeat and SOF domain-containing protein 1; WD repeats and SOF domain containing 1; WD repeats and SOF1 domain containing; WDSOF1
DCAF13
Primary
Affinity chromatography
24GB3745
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6PAC3, Q9NV06
223499, 25879, 362902
A synthesized peptide derived from human DCAF13 (aa 1-46).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.